You can apply filters to the Workflow Presets table to narrow search results, or shorten the list of workflow presets to make it easier to find the workflow presets of interest.
In the Workflows tab, click Presets.
Click the filter category to expand the list, then select a filter from the list.
Option
Description
Annotation Sets
Filter by:
Annotation Sets
Annotation Source
Filter Chains
Copy Number Baselines
Sequence Variant Baselines
Final Report Templates
Fusion Panels
Target Region Files
Hotspot Region Files
Version
Versions of Ion Reporter™ Software.
Reference
Filter by:
GRCh38
hg19
The filtered list of workflow presets is returned in the Workflow Presets table.