You can use keywords or text strings to search for data in the Workflow Presets table.
In the Workflows tab, click Presets.
In Search, enter a search term, then click Go.
The search field is outlined in red if the search string is invalid. The following rules apply to all search fields:
An asterisk (*) is not allowed in the search field.
Searches are not case-sensitive.
Searches match your search string in any location in the target list. For example, a search on "demo" in analysis workflow names matches analysis workflows with "demo" anywhere in their name.
The search results are returned in the Workflow Presets table.