Apply an Analyses table preference
You can apply table preferences to change the view of information in an Analyses table. Table preferences allow you to view information that is relevant to your research in Ion Reporter™ Software.
  1. In the Analyses tab, click Overview.
  2. Click PreferencesTable Preferences.
  3. Select any available table preference that you want to apply to the table.
    The view in the Analyses table changes to show your selected preferences.
  4. (Optional) Create a new Analyses table preference based on the table preference that you selected.
    1. Click PreferencesSelect Columns, select any available columns that you want to add, then click Apply.
    2. Click the column heading that you want to move, then drag and drop the column to the position that you prefer.
    3. Click PreferencesSave Table Preference As.
    4. In the Save Table Preference dialog box, enter a name for the table preference, then click Save.
    The new Analyses table preference is saved.