View a MyVariants database associated with an analysis workflow
You can determine which MyVariants database is selected for an analysis workflow in Ion Reporter™ Software.
  1. In the Workflows tab, click Overview.
  2. Click the row for the analysis workflow of interest.
  3. In the Details pane, look for MyVariants Database.