Default DNA and Fusions View filter
This filter can be added to filter chains in the software to detect all of the fusion variants, expression control markers, assay 5′/3′ markers, and all DNA variants that the predefined Default DNA and Fusions View filter chain detects.
Filter conditions include the ability to select specific annotations (Present or Absent), to include all default DNA and Fusions view variants (In), or to exclude all Default DNA and Fusions view variants (Not In) in filtered analysis results. In addition, unannotated variants can be included or excluded in filtered analysis results.
The filter detects the fusion variants, expression control markers, assay 5′/3′ markers, and all DNA variants, which can be included or excluded in filtered analysis results.