Add attribute columns to the Analysis Results table
You can add columns to the
Analysis Results
table that you view in Ion Reporter™ Software.
Note: In the Summary tab, you can add any of the available columns
from any subtab in the Analysis results table. You can move
or deselect existing columns in the other tabs, such as the Oncomine tab,
Functional tab, Population tab, Ontologies tab, Pharmacogenetics tab, and QC
tab, but cannot add more columns to these tabs.
In the Analyses tab, click Overview.
Click the link for an analysis of interest to open the Analysis Results table.
Click Preferences
> Select Columns, then select the columns that you want to add to the
Analysis Results
table, or deselect the columns that you want to remove from the table.
The selected columns are added to the
Analysis Results
table until you close the table. To save the table with the attributes that you
added, see Create an Analysis Results table preference.