Select
Shannon Diversity for
a view of a spectratyping plot that highlights the Shannon Diversity (entropy) of identified
clones.
This is an example of an IGH spectratyping plot from an Oncomine™ BCR IGH‑LR Assay analysis. Circle color indicates the
Shannon Diversity of clones having a particular variable gene-CDR3 nucleotide length
combination. Portions of the repertoire containing highly expanded clones typically have
a corresponding low Shannon Diversity value. You can further partition the clones by
isotype in Ion Reporter™ Software with the buttons that are above the
interactive spectratyping plot.