Restore default settings for the Analysis Results table
You can restore default column headings in the Analysis Results table Ion Reporter™ Software.
- In the Analyses tab, click Overview.
- Click the link for an analysis of interest to open the Analysis Results table.
- Click .
The default table is restored.