The IonReporterUploader command-line utility is an alternative to the IonReporterUploader plugin that is included with Torrent Suite™ Software.
This procedure explains how to download and extract the IonReporterUploader command-line utility. The procedure can vary, based
on the operating system of the target computer. In general, decompress the
downloaded directory on your target machine, then copy the directory IonReporterUploader-cli to a convenient location.
IMPORTANT! Use only the decompression utility available on your local computer. Do not decompress files on a different operating system and copy those files to a computer that uses a different operating system.
Download the IonReporterUploader command-line utility at: http://iru.ionreporter.thermofisher.com/.
Ideally, download the IonReporterUploader command-line utility onto the computer where it
is to be run. At a minimum, you must use a computer with the same operating
system.
Click the filename IonReporterUploader-cli.zip, then download the file to the target computer.
Extract the downloaded IonReporterUploader-cli.zip file, then copy the IonReporterUploader-cli directory to a convenient location on the target computer.