RefGene GeneModel and Ensembl GeneModel transcript versions
You can view the details about the annotations that Ion Reporter™ Software supports for a particular gene model transcript in the list of Variant Details, including the versions for the RefGene GeneModel and Ensembl GeneModel transcripts.
In the Analysis tab, click Overview.
Click an analysis name to open the Analysis Results screen.
In the Analysis Results screen, click (Variant Details) to open the Variant Details screen.
The transcript version is listed in the Annotation Value column.
Alternatively, open the TSV variants file in the Variants/sample_name subdirectory, to see the transcript versions.
Use these version numbers with the transcript IDs in your preferred transcript file.