View a published final report
You can view a published final report of a completed analysis. A published final report includes details such as location, type, coverage, and annotations. You can download published final reports as PDF files.
A published final report of analysis results cannot be edited. That is, you cannot add annotations, notes, My Variants, or classifications to the analysis. However, you can generate another report if you reanalyze the results.
- In the Analyses tab, click Overview.
- In the Stage column, look for an analysis that is marked as Report Published.
- Select the checkbox for the analysis of interest.
- Click View Final Report.
-
In the report preview, review the report.
You can:
Click Switch To to view a QC report, an audit log, or view analysis results. For more information, see View a Quality Control (QC) report, View and download audit logs and Display different views of analysis results.
×