Spectratyping overview plots are generated for the Oncomine™ BCR IGH‑LR Assay, and are available only in the compressed directory of analysis results.
In the Analyses tab, click Overview.
Click the analysis name hyperlink.
In the Immune Repertoire Results screen, in the Actions column for the analysis of interest, click Download. A ZIP file that contains PNG and PDF files of the plots as well as other results files is downloaded.