Spectratyping overview plot
Spectratyping overview plots highlight the variable gene mutation, evenness, diversity, number of clones, and largest clone frequency for each variable gene-CDR3 nucleotide length combination, with further subdivision of the data by isotype. For the Oncomine™ BCR IGH‑LR Assay analysis workflow, ordering of the isotypes reflects their position within the IGH locus.
Overview plots are generated for all immune repertoire assays used with the analysis workflow, and are available only in the compressed folder of analysis results. For more information, see Download spectratyping overview plots.
Figure 1. IGH V-gene usage and Shannon Diversity spectratyping overview plot