Add a report template to an analysis workflow
If you know which information that you want to include in a final report from sequencing results that are shown in the Analysis Results table, you can add a report template to an Ion Reporter™ Software analysis workflow. Reports published from the analysis results that use the analysis workflow are based on the final report template that is associated with an analysis workflow.
-
Create a custom report template or select a system template.
For more information, see Create a final report template from analysis results or Create a final report template workflow preset.
- In the Workflows tab, click Create.
-
Make the appropriate selections in the Research Application, Reference, Annotation, Filters, Copy Number, and Plugins steps.
For more information, see Create a custom analysis workflow without predefined settings.
- In the Final Report step, under Final Report Template, select the report template that you want to associate with the analysis workflow, then click Next.
-
Make appropriate selections in the
Parameters
step, then enter a Workflow name and optional description in
the Confirm step, then click
Save Workflow.
The report template is now associated with the analysis workflow. When you view the analysis results, you can publish a final report that uses the report template. You can also publish a final report for other analyses that use the analysis workflow with which the report template is associated.