Adjust data and access clone details in spectratyping plots
You can adjust data and access clone details in the interactive spectratyping plots in Ion Reporter™ Software. You can also download a static image of the plot shown in the screen. When you adjust the plot view and download an image of the plot, the adjusted plot is downloaded.
Adjust the view of the data in the plot.
Option
Description
Limit the region (v-genes) that appear in the plot
Drag the ends of the horizontal bar below the X‑axis.