You can sort the Samples table to make it easier to find the samples that you are looking for in Ion Reporter™ Software. You can also sort the table to customize the list of samples. For more information, see Customize the Samples table.
In the Samples tab, in the Overview screen, click a column heading to sort the table based on sample attributes.
Click the column again to reverse the order.
Click the column a third time to stop sorting for the column, then return the column to the order that was used before to the sort.