View software version information
An administrator can view version information about the software packages that are installed and available to users of the Ion Reporter Software. This information is helpful to determine whether a software package is current or whether the package needs to be updated.
  1. In the Admin tab, click Software Versions.
  2. In the Software Versions screen, review the information.
  3. Click Package Version to view the software package installations.
  4. Click Jar Versions to view the JAR installations.