JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. frameshift insertion or deletion Insertion or deletion of the number of nucleotide bases that are not divisible by 3, hence change in reading frame, the grouping of codons, and completely different protein translation from the original. ×