You can create a PDF report of MyVariants in Ion Reporter™ Software.
In the Analysis tab, click Overview.
Click an analysis link to open the Analysis Results screen.
Select the MyVariants that you want to include in the report.
MyVariants are denoted by flags in the Analysis Results screen. You can also use filter chains to filter the analysis results for only flagged variants. For more information, see MyVariants filter .
Click Generate Report. If a report template is associated with the analysis workflow, use the Select Final Report Template dialog box to: