Custom primer sequences for Metagenomics analysis workflows
You can upload a set of primer sequences that were used to prepare your samples for metagenomics analysis workflows. Primer sequences can be uploaded from a FASTA file or entered individually into the software. If primer sequences are provided, the sequences are trimmed from the reads before mapping occurs in the software. The names of forward primers must end in _f* and reverse primer names must end in _r*. Primers in a pair must otherwise have identical names so that the software can match the primers during the analysis.
IMPORTANT! The header for each custom reference sequence must include at least the following information, where sequence is the base-pair sequence:
   >mg|Genus|Species| 
sequence
The header can include the following additional information, if available:
 >mg|Genus|Species|Subspecies/
Strain|Accession#|Kingdom|Phylum|Class|Order|Family|PubMed#|LibraryID#|        
 sequence