Import user-defined variantCaller parameters
If you would like to use user-defined variantCaller plugin variant finding parameters in Ion Reporter™ Software, export the parameters JSON file from Torrent Suite™ Software. Then, import this file from the Parameters screen when creating an analysis workflow.
  1. Export the variantCaller parameters from Torrent Suite™ Software and store the JSON file on your computer or network.
  2. In the Parameters step, select Variant Finding.
  3. Click Select File, then browse to the exported variantCaller parameters JSON file and click Import.
    The variantCaller parameters replace the default settings. Error messages appear if any of the imported parameters are out-of-range or if a JSON file with an incompatible format is imported.