Transcripts in REFSEQ and ENSEMBL canonical transcripts file
In UCSC curated canonical transcripts set, there are some discrepancies when multiple transcripts are used for a given gene. The shorter transcripts have been removed and the transcript with the appris_principal_1 tag is retained in the Ion Reporter™ Software REFSEQ canonical transcript set. This tag is used by gencode to mark the primary transcript of a specific gene. If the appris_principal_1 tag is missing, the longest transcript is used so that there is just one canonical transcript for a given gene. If two transcripts are the same length, Ion Reporter™ Software includes both genes and lists them in alphabetical order.
The UCSC transcripts for the following genes are replaced with the transcripts in the New transcript column in Ion Reporter™ Software:
Gene
Current transcript
New transcript
FGFR2
NM_022970.3
NM_000141.5
FGFR3
NM_001163213.1
NM_000142.5
ABL1
NM_007313.2
NM_005157.6
ARAF
NM_001256196.1
NM_001654.5
CHEK2
NM_001005735.1
NM_007194.4
GNAS
NM_080425.2
NM_000516.7
TP53
NM_001276760.1
NM_000546.6