Check if a sample exists
isSampleExists
URL
/api/v1/isSampleExists
Description
IonReporterUploader plugin checks whether there is an existing sample that uses the sample that is given in the call.
Method
POST
Request query parameters
sampleName
String. Mandatory.
Valid sample name.
Example request
https://ionreporter.thermofisher.com/api/v1/isSampleExists
Request headers
Content-Type:application/x-www-form-urlencoded
Authorization:API Token
Returns
true if sample with given name already exists in the organization given the API token for the user in headers.
false if the sample does not exist.
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -X POST -k https://<server name>/api/v1/isSampleExists -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw" -d "sampleName=Poo_C03-740_DNA_009_think3_IRUCLI"