Create a PValue, dbSNP, and Variant Type filter chain
You can create a custom filter chain with multiple filters in Ion Reporter™ Software. This example shows how to create a filter chain with PValue, dbSNP, and Variant Type filters.
IMPORTANT! If using a single filter with a NOT operator, do not use a parenthesis. If parenthesis is used, the filter chain can be saved but an error occurs when applying the filter chain in analyses results. For example, use NOT Filtered Coverage instead of NOT (Filtered Coverage).
In the Workflows tab, click Presets.
Click Create Preset > Filter Chain.
In the Create Filter Chain dialog box, enter a name and an optional (optional) description.
In the Choose Filter list, select PValue.
Enter a range, then select Include boundary values.