JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Remove filters You can remove individual filters from the Samples table in Ion Reporter™ Software. Remove filters from the Samples table. Click (Remove) next to the filter list to remove the filter from the Samples table. Click More Filters, then click the checkmark of the filter that you want to remove the filter from the Samples table. Click More Filters, then click Clear to remove all the selected filter categories from the list. Click Clear All to remove all filters, then view all samples in the Samples table. Related Topics Parent topic: Find samples ×