Lock a user-defined filter chain in workflow presets
After you create a user-defined filter chain in Ion Reporter™ Software, you can lock it. A locked filter chain cannot be edited or unlocked.
  1. In the Workflows tab, click Presets.
  2. In the search filter category, select Filter Chains.
  3. In the row of the filter chain that you want to lock, click (Actions)Lock.
  4. In the Lock Filter Chain dialog box, click Yes to lock the filter chain.
The filter chain preset has a lock symbol in its row on the Workflow Presets screen.