Get workflow names
getWorkflowNamesWithApplicationTypeWithAuth
URL
/api/v1/getWorkflowNamesWithApplicationTypeWithAuth
Description
Used by IonReporterUploader plugin to get a list of workflows that are tagged with IRU for the given version. Returns the list of workflow details for the version that is specified in the header. If there are no workflows that are tagged with IRU, all workflows for a specified version are returned.
Method
POST
Example request
https://ionreporter.thermofisher.com/api/v1/getWorkflowNamesWithApplicationTypeWithAuth 
Request headers
Content-Type:application/x-www-form-urlencoded
Authorization:API Token
Version: IR52
Returns
[
  {
    "ApplicationType": "Amplicon Sequencing",
    "Workflow": "RK_GSTT1_GRCh38_APR06",
    "irVersion": "52",
    "tag_isFactoryProvidedWorkflow": "false",
    "irReference": "GRCh38"
  }
]
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -X POST -k https://<server name>/api/v1/getWorkflowNamesWithApplicationTypeWithAuth -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw" -H "Version:52"