unsharedAnalyses
|
URL
|
/api/v1/unsharedAnalyses
|
Description
|
Unshares analyses. Returns a JSON file with status and message.
|
Method
|
POST
|
Request query parameters
|
userId
String. Mandatory.
UserId
|
Example request
|
https://ionreporter.thermofisher.com/api/v1/unsharedAnalyses
|
Request headers
|
Content-Type:application/x-www-form-urlencoded
Authorization:API Token
|
Returns
|
{ "status":true, "message":"success"}
|
Response fields
Status code
|
Response message
|
200
|
Successful response
|
400
|
Bad request
|
404
|
Not found
|
401
|
User is not authenticated
|
500
|
Error occurred
|
Curl command
curl -X POST -k "https://<server name>/api/v1/unsharedAnalyses" -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw" -d "userId=ff8081815547ceb401554d62c0ec0493"