Get available cancer types
getAvailableCancerType
URL
/api/v1/getAvailableCancerType
Description
Returns the list of all available cancer types defined in attribute-controlled vocabulary.
Method
POST
Example request
https://ionreporter.thermofisher.com/api/v1/getAvailableCancerType
Request headers
Content-Type:application/x-www-form-urlencoded
Authorization:API Token
Returns
 ["Bladder Cancer","Breast Cancer","Colorectal Cancer","Endometrial Cancer",
"Esophageal Cancer","Gastric Cancer","Gastrointestinal Stromal Tumor","Glioblastoma"
,"Head and Neck Cancer","Kidney Cancer","Liver Cancer","Melanoma","Mesothelioma",
"Non-Small Cell Lung Cancer","Osteosarcoma","Ovarian Cancer","Pancreatic Cancer",
"Prostate Cancer","Skin Basal Cell Carcinoma","Small Cell Lung Cancer",
"Soft Tissue Sarcoma","Testicular Cancer","Thyroid Cancer","Unknown"] 
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -X POST -k https://<server name>/api/v1/getAvailableCancerType -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw"