|
Oncomine™ Comprehensive Fusion Panel
|
EGFRvlll
|
Fusion
|
120
|
Ion PGM™ System/ Ion 318™ Chip
|
Oncomine™ Comprehensive - w2.2 - Fusions
|
|
Oncomine™ Comprehensive - w2.2 - DNA and Fusions
|
|
Oncomine™ Focus Fusion Panel
|
MET exon 14
|
Fusion
|
120
|
Ion PGM™ System/ Ion 318™ Chip
|
Oncomine™ Focus - w2.2 - Fusions
|
|
EGFRvIII
|
Fusion
|
120
|
Oncomine™ Focus - w2.2 - DNA and Fusions
|
|
Oncomine™ Focus Fusion Panel for Ion 520™ Chip
|
MET exon 14
|
Fusion
|
120
|
Ion S5™ System/ Ion 520™ Chip
|
Oncomine™ Focus - 520 -w2.2 - Fusions
|
|
EGFRvIII
|
Fusion
|
120
|
Oncomine™ Focus - 520 - w2.2 - DNA and Fusions
|
|
Oncomine™ Comprehensive & Focus Fusion panel for Ion 540™ Chip
|
MET exon 14
|
Fusion
|
1,000
|
Ion S5™ System/ Ion 540™ Chip
|
Oncomine™ Comprehensive v1 - 540 - w2.2 - DNA and Fusions
|
|
EGFRvIII
|
Fusion
|
1,000
|
Oncomine™ Comprehensive v2 - 540 - w2.2 - DNA and Fusions - Single Sample
|
|
Oncomine™ Comprehensive v3 Panel
|
MET exon 14
|
RNAExon Variant
|
1,000
|
Ion S5™ System/ Ion 540™ Chip
|
Oncomine™ Comprehensive v3 - w3.0 - DNA and Fusions - Single Sample
|
|
EGFRvIII
|
RNAExon Variant
|
1,000
|
Oncomine™ Comprehensive v3 - w3.0 -Fusions - Single Sample
|
|
Others (9 other genes have similar assays)
|
RNAExon Variant
|
Present/ absent calls are not made
|