When you open the Analysis Results table in Ion Reporter™ Software, a default set of columns is included. The table columns are based on various factors, including the samples that are used, analysis and workflow name, and other analysis attributes, and the workflow and annotation set that is applied when the analysis is launched.