Create an Analysis Results table preference
You can create a custom table preference for the Analysis Results table in Ion Reporter™ Software. The table preference allows you to view only the columns of data that are relevant to your research.
The table preference setting is included as output when you download a variants file.
- In the Analyses tab, click Overview.
- Click the link for an analysis of interest to open the Analysis Results table.
-
Click , then select the analysis attributes that you want to add to the
Analysis Results
table, or deselect the analysis attributes that you want to remove from the
table.
The selected columns are added on the right side of the table.
-
To save the custom table as a preference:
Option
Description
.
This setting allows you to name the custom table preference. The preference setting is then selectable from
.This setting becomes the standard view for the user account. To restore default table columns, click
.
×