Acronym for median absolute pairwise difference. Assuming that adjacent amplicons in the genome most likely have the same underlying copy number in a sample, the difference between the log2(read count ratio) values against the reference baseline for all adjacent amplicons contains information for the noise level of the data. The median of the absolute values of all such difference in log2(read count ratio) is the measure for how informative the results fare for copy number estimates.