Share analyses data
You can share analysis data from Ion Reporter™ Software with anyone, whether or not they are users of the software.
Variants cannot be flagged when an analysis report has been shared by a user in another organization. The flag column on the Analysis tab and the option to flag multiple variants are not available for shared analysis reports.
When a share is in progress, the analysis and the samples associated with it cannot be edited, archived, or deleted.
- In the Analyses tab, select the analysis that contains the data that you want to share, then in the analysis Details click .
- In the Share Analysis dialog box, enter the email address of the person with whom you want to share data, then click Share.
A message about the status of the share opens briefly. When a share is in progress, you see this icon:
. You can continue with other tasks in Ion Reporter™ Software while the share is in progress. When the sharing is complete, you and the recipients of the shared data see this icon beside the report that contains shared data:
.
- Click the Home tab, then click Notifications to view notifications about the share.
Actions