JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Q score Phred quality score (Q score) is used to measure the accuracy of the nucleotide sequence generated by the sequencing instrument. The Q score represents the probability that a given base is called incorrectly by the sequencer. ×