Apply an Analysis Results table preference
You can apply an Analysis Results table preference to change the way that you view the information in an Analysis Results table. Table preferences allow you to view information that is relevant to your research in Ion Reporter™ Software.
- In the Analyses tab, click Overview.
- Click the link for an analysis of interest to open the Analysis Results table.
- Click .
-
Select any available Table Preference that you want to apply to the table.
The view in the Analysis Results table changes that are based on the selected table preference.
-
(Optional) Create a new Analysis Results table preference based on the table preference that you selected.
- Click , then select any Available Columns that you want to add, then click Apply.
- Click the column header that you want to move, then drag, then drop it to the position you prefer.
- Click .
- Enter a name for the table preference in the Save Table Preference dialog box, then click Save.
The new Analysis Results table preference is saved.