Edit a filter chain
You can edit a user-defined filter chain to change the conditions or remove filters in Ion Reporter™ Software. You cannot edit a predefined filter chain.
  1. To edit a user-defined filter chain, use one of the following procedures to get started.
    Option
    Description
    Workflow Presets
    1. In the Workflows tab, click Presets.
    2. Select Filter Chains from the list.
    3. Select the row of the filter chain that you want to modify, then click Edit.
    Analysis Results
    1. In the Analyses tab, click Overview.
    2. Click the link of an analysis of interest. The Analysis Results screen appears.
    3. In the right pane, in the Filter Chains section, select the user-defined filter chain that you want to edit, then click (Edit).
    Visualize
    1. In the Analyses tab, click Overview.
    2. Click the link of an analysis of interest. The Analysis Results screen appears.
    3. Click Visualize, then select the IRGV & Generate Report tab.
    4. In the right pane, in the Filter Chains section, select the user-defined filter chain that you want to edit, then click (Edit).
  2. Update the Edit Filter Chain dialog box.
    If you edit a filter chain, and want to cancel the edits, refresh the screen to clear all edits before you click Cancel. Otherwise, some changes that were made in the Edit Filter Chain dialog box, can persist in the dialog box.
    Option
    Description
    Change name
    In Name, change the filter chain name.
    Change description
    In Description, change the filter chain description.
    Change reference
    Under Reference, select GRCh38 or hg 19.
    Modify FilterChain Query
    In the Choose Filter list, select a filter.
    In the FilterChain Query field, cut-and-paste the filter names to change the order in which filters are applied.
    Modify filter conditions
    In the Choose Filter list, select a filter that is included in the filter chain.
    Change the conditions of the filter, then click Add.
    (Optional) Use Update to define and replace the last filter in the list without changing the filter count.
    Delete filters
    To remove a filter that is included in a filter chain, select the filter, then click (Delete).
  3. Click Save.
The edited filter chain is available to be associated as the default filter chain when an analysis workflow is created, or to be used to change an existing default filter chain. For more information, see Add a filter chain to an analysis workflow and Change the default filter chain for an analysis workflow.
It can also be applied to analysis results or a visualization. For more information, see Apply a filter chain to analysis results and Use filter chains to change analysis results.