An administrator can disable an Ion Reporter™ Software user account. A disabled user account can be enabled at a later time.
When a user account is disabled, the email address is not available to be reused for a different user account. If you want to reuse the email address, you must delete the user from the software.
Click the Admin tab.
In the Users screen, click the hyperlinked email address of the user.
In the Edit User dialog box, in the Status list, select Disabled, then click Save.
The user account is disabled. You can enable the account again at any time.