Change the default filter chain for an analysis workflow
Many Ion Reporter™ Software analysis workflows include a
default filter chain. You can change the default filter chain for an analysis
workflow to ensure that the filter chain is applied when the analysis workflow is
launched.
Note: Only one default filter chain can be associated with an analysis workflow.
Click Workflows > Create Workflow.
In the Filters step, select a different filter chain from the list.
Proceed through the remaining steps of the workflow bar, then click Save.
The newly saved filter chain is now the default filter chain for the analysis workflow. All future analyses that are run through the new version of the analysis workflow will use the new filter chain. Analyses that were previously launched with older versions of the analysis workflow are not affected when the default filter chain is changed.