Apply an exon tile fusion baseline workflow preset to an analysis workflow
An exon tile fusion baseline is created from fusion-negative samples to establish a control baseline. For analysis workflows that include RNA Exon Tiling, you can select an exon tile fusion baseline to apply to an analysis workflow.
- In the Workflows tab, start to create or copy an analysis workflow.
-
In the
Exon Tile Fusion Baseline
workflow bar step, select an option from the
Baseline
dropdown list.
Option
Description
If you do not want to identify RNAExon Tiles, select this option.
Select an existing exon tile fusion baseline from the list.
Alternatively, you can click the Workflow Presets link, then configure a new exon tile fusion baseline preset. For more information, see Create an exon tile fusion baseline workflow preset.
- Click Next.
-
Proceed through the remaining steps in the workflow bar.
For more information, see Steps in analysis workflow creation.
- In the Confirm step, click Save Workflow.
×