View sample attributes
When you select a single sample that is listed in the Samples table in Ion Reporter™ Software, you can view details for its attributes in the Details pane. Each column in the table represents a single sample attribute.
  1. In the Samples tab, click Overview.
  2. Select the row that contains the sample of interest.
Details that describe sample attributes, such as gender, role, or type, are listed in the Details pane.