This procedure applies to Ion Reporter™ Server only.
You can import samples that originate in the Ion PGM™ Dx System and are processed by Torrent Suite™ Dx Software to an Ion Reporter™ Server. Only samples that are signed off in Torrent Suite™ Dx Software can be transferred to an Ion Reporter™ Server. The samples are imported in a BAM file.
In the Samples tab, click Overview.
Click Define Samples > Import from PGM™ Dx TS™ Sw.
In the Servers step of the workflow bar, click Add Server.
Enter the name, IP address, user name, and password for the Ion PGM™ Dx System account. Click Save, then click Next.
In the Samples step, select the samples that you want to import, then click Next.
Ensure that the information in the Review step of the workflow bar is correct, then click Import.
The samples appear in the Samples table in Ion Reporter™ Software.
The sample or samples are now ready for analysis. For more information, see Launch an analysis.