Export MyVariants database
You can export a MyVariants database to share it with another user, project, or organization in Ion Reporter™ Software.
  1. In the Analyses tab, click MyVariants.
  2. Click MyVariants, then select Show All, Important, or Ignore.
  3. Click Export TSV.
    A compressed file appears.
  4. When the download is complete, a compressed directory that is named myvariants_results.zip is downloaded in the browser. Save the directory to a hard drive.
  5. Extract the MyVariants directory.
    The TSV file contains the information for each variant, such as Locus, Gene, Type, Flag Type, and Analysis Name. You can also view additional details for each variant such as the list of analyses in which the variant was called.