Confident Somatic Variants filter
This filter detects confident somatic variants in tumor-normal analyses. The filter compares tumor samples to matched normal samples. It does not apply to single sample analyses.
Filter conditions include the ability to include the confident somatic variants (In), or to exclude the confident somatic variants (Not In) in filtered analysis results.
×