This filter can be included in a filter chain in the software to report on MyVariants annotations as set by users.
Filter conditions include the ability to select Important, Ignore, or None for specific MyVariants annotations, and to include or exclude variants that are not flagged in filtered analysis results. Possible combinations include:
|
Option
|
Description
|
|
Important
|
Select Important to filter for MyVariants that are flagged "Important".
|
|
Ignore
|
Select Ignore to filter for MyVariants that are flagged "Ignore".
|
|
Important and Ignore
|
In the Filter Option list, select In to include all flagged MyVariants.
|
|
Ignore and None
|
-
Select Include unannotated variants to include all unflagged variants.
-
Select Ignore to include MyVariants that are flagged "Ignore".
|
|
Important and None
|
-
Select Include unannotated variants to include all unflagged variants.
-
Select Important to include MyVariants that are flagged "Important".
|
|
None
|
In the Filter Option list:
-
Select Not In to not include any flagged MyVariants.
-
Select Include unannotated variants to include all unflagged variants.
|
The filter returns variants in samples that have the specified MyVariants annotations.