A proband variant is assigned the category InconsistentWithFather if all of the
following are true:
-
Neither of the variant's alleles appear in the father's genotype at this
variant position.
-
The variant has coverage at least consistencyMinCoverage.
-
The father's genotype has coverage at least consistencyMinCoverage at this
variant position.