JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Sort the Analyses table Many of the columns in the Analyses table can be sorted to make it easier to find the analysis that you are looking for in Ion Reporter™ Software. In the Analyses tab, in the Overview screen, click a column heading to sort the table based on sample attributes. Click the column heading again to reverse the order. Related Topics Parent topic: Find analyses ×