Create an exon tile fusion baseline workflow
preset
You must use at least 48 fusion-negative RNA
samples to create an exon tile fusion baseline workflow preset.
In the
Workflows tab, click
Presets, then click
Create Preset
> Exon Tile Fusion Baseline.
In the
Baseline Type step of
the
Create Exon Tile Fusion Baseline
workflow bar, select the
Fusion Panel to use for
the exon tile fusion baseline.
Note:
The baseline type is set to
Exon Tile Fusion Baseline
and cannot be changed.
The reference genome is set to
hg19 and cannot
be changed.
Click
Next.
In the
Parameters step, do not
change parameters from the default settings unless you understand how the change can
affect the analysis. Click
Next.
In the
Samples step, select the
samples to be used in the baseline creation.
You must select at least 48 or more fusion-negative RNA samples. If you do not see
samples of interest in the table, see Sample definition for information about how to upload or
define a sample.
Click
Next.
In the
Confirm step, enter a
name, or accept the default name, then enter an optional description for the baseline.
Click
Launch.
The exon tile fusion baseline creation job is started and the baseline with its status now
appears on the screen. When the job completes, the exon tile fusion baseline is added to the
Workflow Presets screen
and is available to add to an analysis workflow. For more information, see Apply an exon tile fusion baseline workflow preset to an analysis workflow.