Analyses table column options
When you open the Analyses table, a default set of columns is included. The table columns are based on various factors, including the samples that are used, the analysis name, the analysis workflow that is used, other analysis attributes, and the analysis workflow and annotation set that is applied when the analysis is launched. You can customize the Analyses table to more easily find analyses of interest, either by the date that the analysis was run, or by sample, analysis workflow, or analysis attributes.
The following columns are included in the Analyses table in Ion Reporter™ Software. Default columns are included for all analyses. You can add other available columns to customize the table.
Column
Description
Default columns—These columns are available by default when you open the Analyses table if no table preferences are applied.
Selection box—Use to select the analysis in the row that contains the selection box. Select one or more analyses to view analysis details, visualize analysis results, or perform other actions on the selected analysis or analyses.
(Checked Out)
Checked out—Indicates that edits to the analysis are in progress. Open the analysis in view-only mode to view analysis results and see the name of the user who is making edits to the analysis.
Analyze role—Indicates that the analysis is shared by a user who has the Analyze role.
 (Add)
Add notes—Use to add a new note to analysis. Analysis notes are time-stamped written notes to capture comments or observations for an analysis that has successfully completed. An analysis can contain multiple notes from different users.
 (Note)
Add/Delete Notes—Use to add a note to analysis, or delete an analysis note.
Analysis
The name of the analysis.
Sample
The name of the sample.
Version
The Ion Reporter™ Software version number of the analysis workflow.
Reference
Indicates the human or other genomic or database reference used in the analysis.
Stage
The stage of the analysis.
Project
The name of the project. Can be inherited from Torrent Suite™ Software.
Workflow
The name of the analysis workflow used in the analysis.
Launched On
The date and time that the analysis was launched.
Status
The status of the analysis.
  • Success
  • Aborted
  • Failed
Available columns — These columns can be added to customize the Analyses table.
Analyzed
Analysis is complete for the samples shown in the Analyses table.
  • True
  • False
Analyzed By
The user name of the person who analyzed the data.
Analyzed On
The date of the analysis.
Annotations
The variant annotations that are added.
Barcode
The name of the specific barcode in the selected barcode kit.
Biopsy Days
The time point post-fertilization at which the sample was taken from an embryo biopsy.
Cancer Type
The type of cancer in the sample.
Cell Num
The cell count of the biopsied material.
Chip ID
The identifying number assigned to the chip.
Chip Type
The chip type that was used when preparing the sample, such as Ion 550™ Chip.
Control Sequencing Kit
The control kit that was used when preparing the sample.
Copy Number Baseline
The set of control samples that were used to create a baseline for detecting copy number variants (CNVs).
Couple ID
The identifier of the couple for use with the Reproductive research application.
Device ID
The identifying number of the sequencing instrument.
Embryo ID
The identifying number or name assigned to an embryo.
Filter Chain
The name of filter chain that was applied.
Fusion Panel
The name of fusion panel that was used.
Gender
Information about the gender of the sample.
Grouping Type
The grouping type.
  • Single
  • Paired
  • Trio
Hotspots Regions
The name of the hotspots file that was used in Torrent Suite™ Software.
Imported By
The name of the user who imported the sample.
Imported On
The date that the sample was imported.
Instrument
The name of the sequencer that was used.
Instrument Type
The type of the sequencer, such as Ion GeneStudio™ S5 Sequencer.
IR Uploader Version
The software version number of the IonReporterUploader plugin that was used in Torrent Suite™ Software.
Launched By
The name of the user who launched the analysis.
Library Kit Name
The name of the library kit that was used in the analysis.
Locked Workflow
Indicates whether the analysis workflow used in the analysis is locked. A locked analysis workflow cannot be edited or unlocked. However, it can be copied.
  • True
  • False
Mouse Strain
The strain of the mouse sample.
MyVariants Database
The name of the MyVariants database that was used.
Percentage Cellularity
The percentage of tumor cells in the sample.
Plan Name
The name of the plan that was used in Torrent Suite™ Software.
Plugin
The name of the plugin or plugins used in the analysis.
Population
The ethnicity indicator for the sample.
Report Generated By
The name of the user who generated the report.
Report Generated On
The date that the report was generated.
Research Application
The research application that applies to the analysis, such as DNA, Aneuploidy, and so on.
Research Category
The research category that applies to the analysis, such as exome, reproductive, oncology, inherited disease, and so on.
Role
The role in a pair or a group of samples, such as mother, father, proband, or unknown.
Run Date
The date of the sequencing run.
Run ID
The identifying number of the sequencing run.
Run Name
The name of the run that was used in Torrent Suite™ Software.
Sample Collection Date
The date that the sample was collected.
Sample ID
(Optional) If you manage samples in an external system (for example, a LIMS), you can enter the identifier from that system in this column.
Sample Preparation Kit
The name of the sample preparation kit used in Torrent Suite™ Software.
Sample Receipt Date
The date that the sample was received.
Sample Transferred
An indication of whether the sample has been transferred to a user in another organization.
  • Transferred—The sample has been transferred to an organization. A sample can be transferred to more than one organization. Click Transfer to see when and to whom the sample was transferred.
  • Available—The sample is available to be transferred.
Sample Type
Information about the relationship between individual samples that are analyzed.
Sequencing Kit Name
The name of the sequencing kit that was used in Torrent Suite™ Software.
Sequencing Run Type
The type of the sequencing run that was used in Torrent Suite™ Software
Tagged for IRU
Indicates whether the analysis workflow used in the analysis is tagged for use with IonReporterUploader plugin.
  • No
  • Yes
Target Regions
The name of the target regions file that was used in Torrent Suite™ Software
Templating Kit Name
The name of the templating kit that was sed in Torrent Suite™ Software.
Torrent Suite Hotspots File
The name of the hotspots file that was used in Torrent Suite™ Software.
Torrent Suite Region File
The name of the target regions file that was used in Torrent Suite™ Software.
Torrent Suite Results Name
The name of the Torrent Suite™ Software run report.
Torrent Suite Version
The version number of the Torrent Suite™ Software that was used for the analysis.
Variant Type Detection
The type of variant cells detected in the analysis.
  • Germline
  • Rare Somatic
  • Somatic
Workflow Revision
The number of the revision of the analysis workflow.