SIFT score
SIFT stands for Sorting Intolerant from Tolerant and is an algorithm for predicting whether an amino acid substitution affects protein function based on sequence homology and the physical properties of amino acids.